Tisk…

Emodin 95% tech.

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone

117751000 117755000
ACRO117751000EA 4270 CZK
ACRO117751000 ACRO117755000
Emodin 95% tech.
Emodin

Appearance: Orange-red Powder

Vzorec: C₁₅H₁₀O₅
MW: 270.24 g/mol
Bod tání: 253 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001207
CAS číslo: 518-82-1

Objednat

Výsledky specifikačního testu

Appearance Orange-red Powder
Infrared spectrum Conforms
HPLC ≥94 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Ammonium (NH4) ≤20 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %
Neutrality Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR