Carboxymethoxylamine hemihydrochloride 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Aminooxyacetic acid hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride, O-(Carboxymethyl)hydroxylamine hemihydrochloride, (Aminooxy)acetic Acid Hemihydrochloride
108430025
108430100
108430500
ACRO108430025EA
4310
CZK
ACRO108430025
ACRO108430100
ACRO108430500
Carboxymethoxylamine hemihydrochloride 98%
Carboxymethoxylamine hemihydrochloride
CAS No.: 2921-14-4
Vzorec:
C₄H₁₁ClN₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00012955 CAS číslo: 2921-14-4 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...