Tisk…

5-Bromo-2-furoic acid 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid

107020250 107021000
ACRO107020250EA 1360 CZK
ACRO107020250 ACRO107021000
5-Bromo-2-furoic acid 99%
5-Bromo-2-furoic acid
Vzorec: C₅H₃BrO₃
MW: 190,98 g/mol
Bod tání: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003239
CAS číslo: 585-70-6

Objednat

Výsledky specifikačního testu

Appearance White to cream to beige to brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 188 to 192 °C
Titration with NaOH ≥98.5 %
Color scale ≤10 APHA
Stabilizer 5 to 30 ppm BHT
Residue after evaporation ≤0.005 %
Carbonyl (as HCHO) ≤0.01 %
Titrable acid ≤0.0016 meq/g
Peroxides ≤0.005 % (H2O2)
Freezing point ≥11.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR