Tisk…

Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 1-Benzyl-3-ethoxycarbonyl-4-piperidone Hydrochloride, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid ethyl ester hydrochloride

105900250 105901000
ACRO105900250EA 2430 CZK
ACRO105900250 ACRO105901000
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 97%
Ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

CAS No.: 1454-53-1

Vzorec: C₁₅H₁₉NO₃
MW: 297,78 g/mol
Bod tání: 160…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00012800
CAS číslo: 1454-53-1

Objednat

Výsledky specifikačního testu

Appearance Cream to pink Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 % (on dry substance)
Water =12 % (K.F.)
Titration with HClO4 ≥96.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR