Tisk…

3-Amino-1-propanesulphonic acid 95%

Dodavatel: Thermo Fisher Scientific
Varování

104430010 104430050
ACRO104430010EA 1640 CZK
ACRO104430010 ACRO104430050
3-Amino-1-propanesulphonic acid 95%
3-Amino-1-propanesulphonic acid

CAS No.: 3687-18-1

Vzorec: C₃H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00008225
CAS číslo: 3687-18-1

Objednat

Výsledky specifikačního testu

Appearance White to light blue Crystalline powder
Infrared spectrum Conforms
Melting point ≥270 °C
Titration with NaOH ≥94.0 %
Phosphate (PO4) ≤3.5 % (by acid extraction)
Calcium (Ca) ≤200 ppm (by acid extraction)
Iron (Fe) ≤150 ppm (by acid extraction)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR