5-Aminolevulinic acid hydrochloride 98+%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
103925000
103920050
ACRO103925000EA
2410
CZK
ACRO103925000
ACRO103920050
5-Aminolevulinic acid hydrochloride 98+%
5-Aminolevulinic acid hydrochloride
CAS No.: 5451-09-2
Vzorec:
C₅H₁₀ClNO₃ MW: 167,59 g/mol Bod tání: 156…158 °C (rozkl.) Storage Temperature: Freezer |
MDL Number:
MFCD00012869 CAS číslo: 5451-09-2 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.0 to 102.0 % |
Loss on drying | ≤2.0 % (105°C, 3 h) (vacuum) |
Sulfated ash | ≤0.5 % |
Solubility | (5 % in water) Clear to slightly hazy colorless to light yellow |
HPLC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...