Tisk…

Vinylbenzyl trimethylammonium chloride 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Flocculant VA-2, Ammonium, trimethyl(p-vinylbenzyl)-, chloride, polymers (8CI), 4-Ethenyl-N,N,N-trimethylbenzenemethanaminium chloride homopolymer

422560250 422561000
ACRO422560250EA 2030 CZK
ACRO422560250 ACRO422561000
Vinylbenzyl trimethylammonium chloride 97%
Vinylbenzyl trimethylammonium chloride

CAS No.: 26616-35-3

Vzorec: C₁₂H₁₈ClN
Bod tání: 240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080690
CAS číslo: 26616-35-3

Objednat

Výsledky specifikačního testu

Appearance White to yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20°C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR