Tisk…

Vanadium(IV) oxide phthalocyanine 85%, Practical grade

Dodavatel: Thermo Scientific
Varování

Synonyms: Vanadyl phthalocyanine

422490250 422491000
ACRO422490250EA 9420 CZK
ACRO422490250 ACRO422491000
Vanadium(IV) oxide phthalocyanine 85%, Practical grade
Vanadium(IV) oxide phthalocyanine

Appearance: Dark blue to purple Powder

Vzorec: C₃₂H₁₆N₈OV
MW: 579,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041966
CAS číslo: 13930-88-6
UN: 3285
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Dark blue to purple Powder
Infrared spectrum Conforms
Assay ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR