Tetrabromophenolphthalein ethyl ester, pure
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ethyl 2-((3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl)benzoate, Chromoionophore VIII
420070010
420070050
420070250
ACRO420070010EA
6990
CZK
ACRO420070010
ACRO420070050
ACRO420070250
Tetrabromophenolphthalein ethyl ester, pure
Tetrabromophenolphthalein ethyl ester
CAS No.: 1176-74-5
Vzorec:
C₂₂H₁₄Br₄O₄ Bod tání: 208…211 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066387 CAS číslo: 1176-74-5 |
Výsledky specifikačního testu
Appearance | Yellow to red Powder |
Infrared spectrum | Conforms |
UV spectrophotometry | ≥80.0 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...