Tisk…

Sulphorhodamine 101 99%, pure laser grade acid-free

Dodavatel: Thermo Scientific

419945000 419940025 419941000
ACRO419945000EA 21290 CZK
ACRO419945000 ACRO419940025 ACRO419941000
Sulphorhodamine 101 99%, pure laser grade acid-free
Sulphorhodamine 101

Appearance: Green Powder

Vzorec: C₃₁H₃₀N₂O₇S₂
MW: 606,708 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012407
CAS číslo: 60311-02-6

Objednat

Výsledky specifikačního testu

Appearance Green Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
UV Lambda max 578 nm (in Methanol)
Fluorescence max 605 nm (in Methanol)
Optimum lasing 614 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥105000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR