Resazurin sodium salt, pure, certified
Dodavatel: Thermo Scientific
|
Varování
|
418900050
418900010
418900250
ACRO418900050EA
3400
CZK
ACRO418900050
ACRO418900010
ACRO418900250
Resazurin sodium salt, pure, certified
Resazurin sodium salt
Vzorec:
C₁₂H₆NNaO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00005036 CAS číslo: 62758-13-8 |
Výsledky specifikačního testu
Appearance | Dark green to black Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥80 % |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...