Polyvinylsulphuric acid potassium salt, M.W. ∼ 175,000
Dodavatel: Thermo Scientific
|
Varování
|
418140050
418140250
418141000
ACRO418140050EA
9050
CZK
ACRO418140050
ACRO418140250
ACRO418141000
Polyvinylsulphuric acid potassium salt, M.W. ∼ 175,000
Polyvinylsulphuric acid potassium salt
CAS No.: 26837-42-3
Vzorec:
(C₂H₃KO₄S)n MW: 162,2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00081923 CAS číslo: 26837-42-3 |
Výsledky specifikačního testu
Appearance | Off-white to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥21.2 % (K) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...