Tisk…

Biphenyl diphenyl ether eutectic

Dodavatel: Thermo Scientific
Varování

Synonyms: Dowtherm® A, Diphyl

417521000 417520010
ACRO417521000EA 1130 CZK
ACRO417521000 ACRO417520010
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic

Appearance: Clear colorless to yellow Liquid

Vzorec: C₂₄H₂₀O
MW: 324,423 g/mol
Bod tání: 13 °C
Hustota: 1,056 g/cm³
Bod vzplanutí: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00148859
CAS číslo: 8004-13-5
UN: 3082
ADR: 9,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5885 to 1.5905 (20°C, 589 nm)
GC mix of phenyl ether-biphenyl: approx 70%-30%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR