Potassium methyl sulphate 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Methylsulphuric acid potassium salt
414891000
414890100
ACRO414891000EA
10630
CZK
ACRO414891000
ACRO414890100
Potassium methyl sulphate 99%
Potassium methyl sulphate
Appearance: White Crystalline powder or flakes
Vzorec:
CH₃KO₄S MW: 150.189 g/mol Bod tání: 212…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040472 CAS číslo: 562-54-9 UN: 2811 ADR: 6.1 II |
Výsledky specifikačního testu
Appearance | White to almost white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Titration after Ion exchange | ≥98.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...