Indocyanine green, pure laser grade
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen
412541000
412545000
412540020
ACRO412541000EA
3680
CZK
ACRO412541000
ACRO412545000
ACRO412540020
Indocyanine green, pure laser grade
Indocyanine green
Vzorec:
C₄₃H₄₇N₂NaO₆S₂ MW: 774,99 g/mol Bod tání: 235 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013078 CAS číslo: 3599-32-4 |
Výsledky specifikačního testu
Appearance | Green to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
UV Lambda max | 784 nm (in Methanol) |
Molar absorption coefficient | ≥230000 l/(mol.cm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...