Tisk…

Hydrazine acetate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

411750250 411750050
ACRO411750250EA 6550 CZK
ACRO411750250 ACRO411750050
Hydrazine acetate 97%
Hydrazine acetate

Appearance: White to yellow Adhering powder or crystals

Vzorec: H₂NNH₂·CH₃CO₂H
Bod tání: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00013141
CAS číslo: 7335-65-1
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to yellow Adhering powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR