Tisk…

Heparin sodium salt

Dodavatel: Thermo Scientific

Synonyms: Reviparin sodium

411210010 411210050 411212500
ACRO411210010EA 5370 CZK
ACRO411210010 ACRO411210050 ACRO411212500
Heparin sodium salt
Heparin sodium salt

Appearance: White to almost white Powder

Storage Temperature: Ambient
MDL Number: MFCD00081689
CAS číslo: 9041-08-1

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Assay ≥180 IU/mg (dried substance)
Loss on drying ≤8.0 % (1 g, 60°C, 3 h) (P2O5)
UV at 260 nm A: ≤0.15
Appearance of solution faint yellow
NMR (1H-NMR) Chondroitin sulfate (over-sulfated): Absent
Protein ≤0.5 % (dried substance)
pH 5.5 to 8.0 (0.1g in 10ml water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR