Tisk…

Glyoxylic acid 50% in water

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Oxoacetic acid

411060050 411065000 411060025
ACRO411060050EA 802 CZK
ACRO411060050 ACRO411065000 ACRO411060025
Glyoxylic acid 50% in water
Glyoxylic acid
Vzorec: C₂H₂O₃
MW: 74.04 g/mol
Bod tání: –93 °C
Storage Temperature: Ambient
MDL Number: MFCD00006958
CAS číslo: 298-12-4
UN: 3265
ADR: 8 III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Solution
Refractive index 1.4130 to 1.4200 (20°C, 589 nm)
Titration with NaOH ≥50 %
Enantiomeric excess ≥97.5 % (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR