Diethyl oxalacetate sodium salt 95% pract.
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Sodium diethyl oxalacetate, Sodium Diethyl Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt
407371000
407375000
ACRO407371000EA
1520
CZK
ACRO407371000
ACRO407375000
Diethyl oxalacetate sodium salt 95% pract.
Diethyl oxalacetate sodium salt
CAS No.: 40876-98-0
Vzorec:
C₈H₁₂O₅ MW: 210.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00035571 CAS číslo: 40876-98-0 |
Výsledky specifikačního testu
Appearance | Beige to light orange Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...