Tisk…

Zinc(II) dibenzyldithiocarbamate 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: Dibenzyldithiocarbamic acid zinc salt, Dibenzyldithiocarbamic acid Zinc(II) salt, Zinc dibenzyldithiocarbamate

406440250 406441000
ACRO406440250EA 5810 CZK
ACRO406440250 ACRO406441000
Zinc(II) dibenzyldithiocarbamate 95%
Zinc(II) dibenzyldithiocarbamate

Appearance: White to off-white Powder

Vzorec: C₁₅H₁₅NS₂
MW: 610,19 g/mol
Bod tání: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00014470
CAS číslo: 14726-36-4
UN: 3077
ADR: 9,III

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 183 to 188 °C
Assay ≥94 % (ex Zn)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR