Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Di-μ-hydroxo-bis[(N,N,N'.N'-tetramethylethylenediamine)copper(II)] chloride
348980050
348980250
ACRO348980050EA
2010
CZK
ACRO348980050
ACRO348980250
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 98%
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride
Appearance: Dark purple Powder
Vzorec:
C₁₂H₃₄Cl₂Cu₂N₄O₂ MW: 464.42 g/mol Bod tání: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321163 CAS číslo: 30698-64-7 |
Výsledky specifikačního testu
Appearance | Dark purple Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...