Tisk…

1,2-O-Isopropylidene-ɑ-D-xylofuranose 99%

Dodavatel: Thermo Scientific

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

348880100 348880250 348881000
ACRO348880100EA 4710 CZK
ACRO348880100 ACRO348880250 ACRO348881000
1,2-O-Isopropylidene-ɑ-D-xylofuranose 99%
1,2-O-Isopropylidene-alpha-D-xylofuranose

Appearance: White Waxy solid or coarse crystals and/or chunks

Vzorec: C₈H₁₄O₅
MW: 190,20 g/mol
Bod varu: 112…114 °C (,1 mmHg)
Bod tání: 68…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063295
CAS číslo: 20031-21-4

Objednat

Výsledky specifikačního testu

Appearance White Waxy solid or coarse crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation -17° to -19° (20°C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR