2',7'-Dichlorofluorescein 3',6'-diacetate 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2',7'-Dichlorofluorescin diacetate, 2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester, 2',7'-Dichloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate, H₂DCFDA
347250010
347250050
ACRO347250010EA
2810
CZK
ACRO347250010
ACRO347250050
2',7'-Dichlorofluorescein 3',6'-diacetate 97%
2',7'-Dichlorofluorescein 3',6'-diacetate
CAS No.: 2044-85-1
Appearance: Colorless to off-white Powder or crystals
Vzorec:
C₂₄H₁₄Cl₂O₇ MW: 485,269 g/mol Bod tání: 239…246 °C Bod vzplanutí: >109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037501 CAS číslo: 2044-85-1 UN: 1759 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Colorless to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 239 to 246 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...