Flavin adenine dinucleotide disodium salt
Dodavatel: Thermo Scientific
Synonyms:
(((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl)methoxy)(((((2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl)oxy)(hydroxy)phosphoryl)oxy)phosphinic acid Disodium Salt, FAD-Na2, Flavin Adenine Dinucleotide Disodium Salt, Riboflavin 5'-adenosine diphosphate disodium salt, FAD Disodium Salt
347212500
347210010
ACRO347212500EA
4640
CZK
ACRO347212500
ACRO347210010
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
CAS No.: 84366-81-4
Vzorec:
C₂₇H₃₃N₉O₁₅P₂ MW: 829,52 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00151217 CAS číslo: 84366-81-4 |
Výsledky specifikačního testu
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
Water | ≤10 % (K.F.) |
UV spectrophotometry | molar absorptivity 11300 +/- 4%, on dry substance |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...