Tisk…

α-D-Galactose-1-phosphate dipotassium salt pentahydrate 98%

Dodavatel: Thermo Fisher Scientific

337035000 337030025 337030500
ACRO337035000EA 15300 CZK
ACRO337035000 ACRO337030025 ACRO337030500
α-D-Galactose-1-phosphate dipotassium salt pentahydrate 98%
alpha-D-Galactose-1-phosphate dipotassium salt pentahydrate

CAS No.: 19046-60-7

Vzorec: C₆H₁₅K₂O₁₁P
Storage Temperature: Freezer
MDL Number: MFCD00150979
CAS číslo: 19046-60-7

Objednat

Výsledky specifikačního testu

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤23 % (K.F.)
Solubility (5 % in water) Clear colorless
Specific optical rotation +81° to +84° (20°C, 589 nm) (c=2, H2O)
Titration with HClO4 ≥97.5 %
Phosphorus (P) ≤0.2 % (P inorganic)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR