1,2,2,6,6-Pentamethylpiperidin-4-amine 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Amino-1,2,2,6,6-pentamethylpiperidine
335940010
335940050
ACRO335940010EA
1820
CZK
ACRO335940010
ACRO335940050
1,2,2,6,6-Pentamethylpiperidin-4-amine 99%
1,2,2,6,6-Pentamethylpiperidin-4-amine
Appearance: Clear colorless to yellow Liquid
Vzorec:
C₁₀H₂₂N₂ Bod varu: 215…218 °C (760 mmHg) Bod vzplanutí: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861832 CAS číslo: 40327-96-6 UN: 2735 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...