Diethylphosphonoacetic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
330670050
330670250
ACRO330670050EA
1140
CZK
ACRO330670050
ACRO330670250
Diethylphosphonoacetic acid 98%
Diethylphosphonoacetic acid
Appearance: Clear colorless to yellow Viscous liquid
Vzorec:
C₆H₁₃O₅P Bod varu: 150 °C Hustota: 1,22 g/cm³ Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192032 CAS číslo: 3095-95-2 |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4440 to 1.4460 (20°C, 589 nm) |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...