Tisk…

3-(2-Furyl)propanoic acid 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-(Furan-2-yl)propanoic acid

330100010 330100100
ACRO330100010EA 1790 CZK
ACRO330100010 ACRO330100100
3-(2-Furyl)propanoic acid 98%
3-(2-Furyl)propanoic acid

Appearance: White to brown Crystalline powder

Vzorec: C₇H₈O₃
MW: 140,138 g/mol
Bod tání: 56…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005346
CAS číslo: 935-13-7

Objednat

Výsledky specifikačního testu

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR