Tisk…

N-Methyl-D-aspartic acid 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid

329190500 329192500
ACRO329190500EA 2270 CZK
ACRO329190500 ACRO329192500
N-Methyl-D-aspartic acid 98%
N-Methyl-D-aspartic acid

CAS No.: 6384-92-5

Vzorec: C₅H₉NO₄
MW: 147.13 g/mol
Bod tání: 187…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004226
CAS číslo: 6384-92-5

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -26° to -30° (20°C, 589 nm) (c=1, 6 N HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR