Tisk…

N-Methyl-N-octylamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

325620250 325621000
ACRO325620250EA 6110 CZK
ACRO325620250 ACRO325621000
N-Methyl-N-octylamine 98%
N-Methyl-N-octylamine

Appearance: Clear colorless to yellow Liquid

Vzorec: C₉H₂₁N
Bod varu: 68 °C (8 mmHg)
Hustota: 0,79 g/cm³
Bod vzplanutí: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00048927
CAS číslo: 2439-54-5
UN: 2735
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4290 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR