Tisk…

tert-Butylphosphonic acid 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

321520010 321520050
ACRO321520010EA 1910 CZK
ACRO321520010 ACRO321520050
tert-Butylphosphonic acid 98%
tert-Butylphosphonic acid

CAS No.: 4923-84-6

Vzorec: C₄H₁₁O₃P
Bod tání: 188…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00012296
CAS číslo: 4923-84-6
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 192 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR