Tisk…

(Triphenylphosphoranylidene)acetaldehyde 97%

Dodavatel: Thermo Fisher Scientific
Varování

307490050 307490250 307491000
ACRO307490050EA 2120 CZK
ACRO307490050 ACRO307490250 ACRO307491000
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde

CAS No.: 2136-75-6

Vzorec: C₂₀H₁₇OP
Bod tání: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006994
CAS číslo: 2136-75-6

Objednat

Výsledky specifikačního testu

Appearance Cream to pink to orange to brown Crystalline powder
Infrared spectrum Conforms
Water ≤4.0 % (K.F.)
Titration with HClO4 96.0 to 104.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR