Tisk…

2-Chloro-4-hydroxybenzoic acid hydrate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Carboxy-3-chlorophenol hydrate

304200010 304200050
ACRO304200010EA 2360 CZK
ACRO304200010 ACRO304200050
2-Chloro-4-hydroxybenzoic acid hydrate 98%
2-Chloro-4-hydroxybenzoic acid hydrate
Vzorec: C₇H₅ClO₃·xH₂O
MW: 172,57 g/mol (anhy)
Bod varu: 350 °C (1013 hPa)
Bod tání: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00798116
CAS číslo: 56363-84-9

Objednat

Výsledky specifikačního testu

Appearance Off-white to beige to orange-brown Crystalline powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Titration with TBAH ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR