Sodium metatungstate hydrate, crystalline
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
304090500
304092500
304090010
ACRO304090500EA
3050
CZK
ACRO304090500
ACRO304092500
ACRO304090010
Sodium metatungstate hydrate, crystalline
Sodium metatungstate hydrate
CAS No.: 652154-12-6
Vzorec:
H₂Na₆O₄₀W₁₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150663 CAS číslo: 652154-12-6 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder or beads |
Assay | ≥85 % (WO3) |
Stabilizer | ≤200 ppm (Cr-salt) Typical |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...