Tisk…

Penta-O-acetyl-β-D-glucopyranose 98%

Dodavatel: Thermo Scientific

Synonyms: β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

302961000 302965000
ACRO302961000EA 2430 CZK
ACRO302961000 ACRO302965000
Penta-O-acetyl-β-D-glucopyranose 98%
Penta-O-acetyl-beta-D-glucopyranose

CAS No.: 604-69-3

Vzorec: C₁₆H₂₂O₁₁
MW: 390,34 g/mol
Bod tání: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00006597
CAS číslo: 604-69-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.5 %
GC ≥97.5 %
Specific optical rotation +4° to +6° (20°C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR