Methyl-1-hydroxy-1-cyclopropane carboxylate 90%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 1-hydroxycyclopropanecarboxylate
302110010
302110050
ACRO302110010EA
4930
CZK
ACRO302110010
ACRO302110050
Methyl-1-hydroxy-1-cyclopropane carboxylate 90%
Methyl-1-hydroxy-1-cyclopropane carboxylate
Appearance: Clear yellow Liquid
Vzorec:
C₅H₈O₃ MW: 116,116 g/mol Bod varu: 70 °C (13 mmHg) Bod vzplanutí: 17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093884 CAS číslo: 33689-29-1 UN: 3272 ADR: 3,II |
Výsledky specifikačního testu
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...