6-Chloro-2-benzoxazolethiol 99%
Dodavatel: Thermo Scientific
Synonyms:
6-Chlorobenzo[d]oxazole-2(3H)-thione
297300050
297300250
ACRO297300050EA
868
CZK
ACRO297300050
ACRO297300250
6-Chloro-2-benzoxazolethiol 99%
6-Chloro-2-benzoxazolethiol
Appearance: Yellow to beige Crystalline powder or needles
Vzorec:
C₇H₄ClNOS MW: 185,625 g/mol Bod tání: 229…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800495 CAS číslo: 22876-20-6 |
Výsledky specifikačního testu
Appearance | Yellow to beige Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 229 to 232 °C |
HPLC | ≥98.5 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...