Tisk…

Diphenyl-N-cyanocarbonimidate 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Diphenyl N-cyanocarbonimidate, N-Cyanocarbonimidic Acid Diphenyl Ester

297210100 297210500
ACRO297210100EA 3440 CZK
ACRO297210100 ACRO297210500
Diphenyl-N-cyanocarbonimidate 97%
Diphenyl-N-cyanocarbonimidate

CAS No.: 79463-77-7

Vzorec: C₁₄H₁₀N₂O₂
MW: 238,25 g/mol
Bod tání: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00010380
CAS číslo: 79463-77-7

Objednat

Výsledky specifikačního testu

Appearance White Powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
Assay ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR