Tisk…

(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

296420025 296420100 296420500
ACRO296420025EA 2620 CZK
ACRO296420025 ACRO296420100 ACRO296420500
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate

CAS No.: 115314-14-2

Vzorec: C₉H₉NO₆S
MW: 259,23 g/mol
Bod tání: 61…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064582
CAS číslo: 115314-14-2

Objednat

Výsledky specifikačního testu

Appearance Almost white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 61 to 65 °C
GC ≥97.5 %
Specific optical rotation +21.0° to +25.0° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR