Tisk…

(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methyl N-(Triethylammoniosulphonyl)carbamate, Burgess Reagent

294510010 294510050 294510250
ACRO294510010EA 1850 CZK
ACRO294510010 ACRO294510050 ACRO294510250
(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt 97%
(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt

CAS No.: 29684-56-8

Vzorec: C₈H₁₈N₂O₄S
MW: 238.30 g/mol
Bod tání: 76…79 °C
Storage Temperature: Freezer
MDL Number: MFCD00077815
CAS číslo: 29684-56-8

Objednat

Výsledky specifikačního testu

Appearance White to yellow to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR