Tisk…

(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine 96%

Dodavatel: Thermo Scientific

293462500 293460010
ACRO293462500EA 5890 CZK
ACRO293462500 ACRO293460010
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine 96%
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine

CAS No.: 93621-94-4

Vzorec: C₈H₁₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00210016
CAS číslo: 93621-94-4

Objednat

Výsledky specifikačního testu

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4500 (20°C, 589 nm)
GC ≥95 %
Specific optical rotation +7° to +9° (20°C, 589 nm) (c=3, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR