Tisk…

Isobornyl methacrylate 85-90% stabilised

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate, (1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate(relative), Methacrylic acid Isobornyl Ester

292092500 292090010
ACRO292092500EA 2730 CZK
ACRO292092500 ACRO292090010
Isobornyl methacrylate 85-90% stabilised
Isobornyl methacrylate

CAS No.: 7534-94-3

Vzorec: C₁₄H₂₂O₂
MW: 222.33 g/mol
Bod varu: 245 °C (760 mmHg)
Bod tání: –60 °C
Hustota: 0.98 g/cm³
Bod vzplanutí: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00081070
CAS číslo: 7534-94-3

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
GC ≥85 %
Stabilizer 100 to 200 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR