Dicobalt octacarbonyl 95% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Di-Cobalt Octacarbonyl, Cobalt carbonyl (Dicobalt octacarbonyl), Octacarbonyldicobalt, Cobalt Carbonyl, Cobalt carbonyl
291840050
291840250
291841000
ACRO291840050EA
2250
CZK
ACRO291840050
ACRO291840250
ACRO291841000
Dicobalt octacarbonyl 95% stabilised
Dicobalt octacarbonyl
CAS No.: 10210-68-1
Vzorec:
C₈Co₂O₈ MW: 341.95 g/mol Bod tání: 51…52 °C (rozkl.) Hustota: 1.81 g/cm³ Bod vzplanutí: –13 °C Storage Temperature: Freezer |
MDL Number:
MFCD00016024 CAS číslo: 10210-68-1 UN: 3124 ADR: 6.1 I |
Výsledky specifikačního testu
Appearance | Orange-red to dark red Crystals or flakes |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Stabilizer | ≤5 % hexane |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...