Acetylsalicylsalicylic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
269352500
269350010
ACRO269352500EA
2800
CZK
ACRO269352500
ACRO269350010
Acetylsalicylsalicylic acid 97%
Acetylsalicylsalicylic acid
CAS No.: 530-75-6
Vzorec:
C₁₆H₁₂O₆ Bod tání: 145…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143537 CAS číslo: 530-75-6 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 145 to 160 °C |
Water | ≤0.5 % |
HPLC | ≥96.0 % (As is) Calculated |
Titration with NaOH | ≥96.0 % (On anhydrous substance) |
Residual solvents | ≤2.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...