Tisk…

2-Phenyl-5-benzimidazolesulphonic acid 98%

Dodavatel: Thermo Scientific

Synonyms: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

259700250 259700050 259702500
ACRO259700250EA 1490 CZK
ACRO259700250 ACRO259700050 ACRO259702500
2-Phenyl-5-benzimidazolesulphonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid

CAS No.: 27503-81-7

Vzorec: C₁₃H₁₀N₂O₃S
MW: 274,29 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00053007
CAS číslo: 27503-81-7

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Titration excess NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR