Ethyl-2-(bromomethyl)acrylate 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ethyl 2-(bromomethyl)acrylate
235360050
235360010
ACRO235360050EA
8760
CZK
ACRO235360050
ACRO235360010
Ethyl-2-(bromomethyl)acrylate 97%
Ethyl-2-(bromomethyl)acrylate
Appearance: Clear colorless to light yellow Liquid
Vzorec:
C₆H₉BrO₂ Bod varu: 86 °C (20 mmHg) Hustota: 1,38 g/cm³ Bod vzplanutí: 81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00031518 CAS číslo: 17435-72-2 UN: 3265 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4810 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...