Tisk…

Folinic acid calcium salt pentahydrate 95.0-105.0%

Dodavatel: Thermo Fisher Scientific

230310010 230312500
ACRO230310010EA 6330 CZK
ACRO230310010 ACRO230312500
Folinic acid calcium salt pentahydrate 95.0-105.0%
Folinic acid calcium salt pentahydrate

Appearance: White to yellow Powder

Vzorec: C₂₀H₃₁CaN₇O₁₂
Bod tání: 240…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149465
CAS číslo: 6035-45-6

Objednat

Výsledky specifikačního testu

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤17 %
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR