N-Cbz-L-serine 99+%
Dodavatel: Thermo Scientific
Synonyms:
N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine
229510050
229510250
ACRO229510050EA
1110
CZK
ACRO229510050
ACRO229510250
N-Cbz-L-serine 99+%
N-Cbz-L-serine
Appearance: White to cream Powder
Vzorec:
C₁₁H₁₃NO₅ MW: 239,2 g/mol Bod tání: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002662 CAS číslo: 1145-80-8 |
Výsledky specifikačního testu
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | 115 to 120 °C |
Water | ≤0.5 % (K.F.) |
HPLC | ≥99.0 % |
Specific optical rotation | +5.5° to +6.5° (20 °C, 589 nm) (c=7, AcOH) |
Titration with NaOH | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...