Tisk…

4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: GLCNAC1-B-PNP, 4-Nitrophenyl N-Acetyl-β-D-glucosaminide, 4-Nitrophenyl 2-acetamido-2-deoxy-beta-D-glucopyranoside

229411000 229410010
ACRO229411000EA 1560 CZK
ACRO229411000 ACRO229410010
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside 99%
4-Nitrophenyl-2-acetamido-2-deoxy-beta-D-glucopyranoside

CAS No.: 3459-18-5

Vzorec: C₁₄H₁₈N₂O₈
MW: 342.30 g/mol
Bod tání: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00063696
CAS číslo: 3459-18-5

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (0.5 % in water) Clear colorless to light yellow
HPLC ≥98.5 %
Specific optical rotation -13° to -17° (20°C, 589 nm) (c=0.5, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR