Methyl-ɑ-D-mannopyranoside 99+%
Dodavatel: Thermo Fisher Scientific
Synonyms:
ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
229251000
229250250
229255000
ACRO229251000EA
4750
CZK
ACRO229251000
ACRO229250250
ACRO229255000
Methyl-ɑ-D-mannopyranoside 99+%
Methyl-alpha-D-mannopyranoside
CAS No.: 617-04-9
Vzorec:
C₇H₁₄O₆ Bod tání: 186…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063262 CAS číslo: 617-04-9 |
Výsledky specifikačního testu
Appearance | White to off-white to light grey Powder |
Infrared spectrum | Conforms |
Melting point | 186 to 196 °C |
Water | ≤0.5 % |
Sulfated ash | ≤0.2 % |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | +77° to +81° (20 °C, 589 nm) (c=10, water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...